Learn More
Thermo Scientific™ 11mm Plastic Crimp Top Autosampler MicroVials, 250µL virgin polypropylene
Choose from these chemically inert polypropylene 11mm vials that are suitable for most chromatography applications.
Brand:  Thermo Scientific™ C4011-13
Code : 50
Additional Details : Weight : 0.13500kg
Product Code. 11588240
  • 18.95 € / Pack of 100
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
Choose from these chemically inert polypropylene vials,  manufactured from high-quality, GC-tested polypropylene material, that are suitable for most chromatography applications. Thermo Scientific™ 11mm Plastic Crimp/Snap Top Vials provide an economical alternative to glass for some applications. Polypropylene is chemically inert and suitable for most chromatography applications, and can be used when levels of inorganic ions have to be kept to an absolute minimum. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
  • Constructed of polypropylene
  • Precision formed tips are designed to aid sample recovery
  • 12 × 32mm vials
  • Compatible with aluminum seals or Snap-It™ snap cap closures