Learn More
Cytiva Fetal Bovine Serum, Heat Inactivated
GE Hyclone Fetal Bovine Serum, Heat Inactivated, is sourced from South America and complies with the EU regulations.
Brand: Cytiva SV30160.03HI
256.01 EUR valid until 2024-12-31
Use promo code "21878" to get your promotional price.
Code : 85
Additional Details : Weight : 0.54000kg
Description
- Filtered through 3 sequential 100μm pore-size rated filters
- Full traceability compliant with current Good Manufacturing Practices
- Extensive quality, performance and analytical testing
- Aseptically packaged in a Class 100 cleanroom
- Each lot of serum is tested to ensure quality and sterility
- Pooled after filtration and before final packing
- Batch testing available on request
- Suitable for research applications
- Provided in a 500mL bottle
Specifications
South America | |
500 mL | |
Yes |
Fetal Bovine Serum | |
Research applications |