missing translation for 'onlineSavingsMsg'
Learn More

Surface Decontaminant DNA AWAY, Bottle 4L

Brand:  Mbp Molecular Bio Products 7012-11

Surface Decontaminant DNA AWAY, Bottle 4L

Code : NEW

Additional Details : Weight : 3.78000kg

Product Code. 15676368

  • 146.00 € / 4 L
In Stock
to see stock.
Add to basket