missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius TSA Nutrient Agar in Bottles
TSA Nutrient Agar media in Bottles for colony counting by the membrane filtration method or via direct incubation.
Brand: Sartorius 14171----------A
Additional Details : Weight : 3.00000kg
Description
- Time- and cost-saving
- Compliance with international standards and references
- Consistent quality
- The traditional culture media for microorganisms is agar media.
- The media are available in two formats.
- Agar media in tubes are for pouring agar plates.
- The content of one tube is sufficient for two 90 mm or three 60 mm petri dishes.
- Agar media in bottles are the cost-effective alternative for casting plates.
- Agar media in tubes are for pouring agar plates.
Specifications
TSA Nutrient Agar | |
Sterile | |
Bottle |