missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas

Manufactured from stainless steel. Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas are multi-purpose.

Brand:  Bochem™ 3018

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10431571

  • 2.35 € / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

  • Double endet
  • Stainless steel
  • Autoclavable
  • Straight
Specifications

Specifications

1 Ea.
Straight
Multi-purpose spatula for a wide range of uses
4 mm
Stainless Steel (18/10)
150 x 4 mm
150 mm
Autoclavable
Double Ended Micro Spatula
40 mm
4 mm
2mm
Stainless Steel (18/10)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers