missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Double Spatulas, Round/Bent Ends, Type 1

Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Double Spatulas serve many uses in the lab.

Brand:  Bochem™ 3182

Code : A0

Additional Details : Weight : 0.00630kg

 View more versions of this product

Product Code. 10028300

  • 2.80 € / Each

In Stock
to see stock.
Add to basket

Description

Description

  • Double spatula
  • Manufactured from 18/10 stainless steel
Specifications

Specifications

1 Ea.
Double Ended Spatula
5 mm
Stainless Steel (18/10)
Round/Bent Ends, Type 1
12 mm
5 mm
210 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers