missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ 18/10 Stainless Steel Double Spatulas, Round/Bent Ends, Type 1
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Double Spatulas serve many uses in the lab.
Brand: Bochem™ 3180
Code : A0
Additional Details : Weight : 0.00090kg
Description
- Double spatula
- Manufactured from 18/10 stainless steel
Specifications
1 Ea. | |
Double Ended Spatula | |
5 mm | |
Stainless Steel (18/10) |
Round/Bent Ends, Type 1 | |
10 mm | |
5 mm | |
130 mm |