missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Laboratory Spoons

Flat-handled, oval-bowled spoons constructed of stainless steel. Bochem™ 18/10 Stainless Steel Laboratory Spoons are chemically resistant, sturdy and polished for easy cleaning.

Brand:  Bochem™ 3391

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10418242

  • 5.55 € / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

  • Durable 18/10 stainless steel construction.
  • Polished surface for easy cleaning.
  • Oval bowl for easy material pickup and delivery.
Specifications

Specifications

1 Ea.
Laboratory Spoon
10mm
200 mm
Autoclavable
Multi-purpose spoon for a wide range of uses
Stainless Steel (18/10)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers