Learn More
Bochem™ 18/10 Stainless Steel Laboratory Spoons
Flat-handled, oval-bowled spoons constructed of stainless steel. Bochem™ 18/10 Stainless Steel Laboratory Spoons are chemically resistant, sturdy and polished for easy cleaning.
Brand:  Bochem™ 3390
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10564261
  • 4.95 € / Each
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
Description
Description
  • Durable 18/10 stainless steel construction.
  • Polished surface for easy cleaning.
  • Oval bowl for easy material pickup and delivery.
Specifications
Specifications
Autoclavable
Multi-purpose spoon for a wide range of uses
Stainless Steel (18/10)
Laboratory Spoon
10mm
150 mm