Learn More
IBA Lifesciences Strep-Tactin™ Sepharose™ Column
Strep-Tactin™ Sepharose™ column is a Gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
88.00 € - 188.50 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12886436
|
IBA Lifesciences
2-1202-001 |
1 mL |
88.00 €
1mL |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
12836446
|
IBA Lifesciences
2-1202-550 |
0.2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Strep-Tactin™ Sepharose™ columns contains 4% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- Due to the low concentrated agarose, it is particularly suitable for large proteins, especially if the affinity to Strep-Tactin™XT 4Flow™ is too high.
- The wide-meshed polymer allows for the simple attachment of larger proteins to the stationary phase.
- To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
For Research Use Only.