missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-3-Aminocyclopentanecarboxylic acid, 95%, 98% ee, ACROS Organics™

Brand:  Acros Organics 362270050

Code : 29

Additional Details : CAS Number : 71830-08-5 Weight : 0.05500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 11955701

  • 1714.00 € / 5g

Estimated Shipment: 08-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

71830-08-5
129.16
1r,3s-3-aminocyclopentanecarboxylic acid, 1r,3s-3-aminocyclopentane-1-carboxylic acid, --1r,3s-3-aminocyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1r,3s, 1r,3s---3-aminocyclopentanecarboxylic acid, cis-3-amino-cyclopentanecarboxylic acid, 1r,3s-3-amino-cyclopentanecarboxylic acid, pubchem18731, 1r,3s---3-amino-cyclopentanecarboxylic acid
(1R,3S)-3-aminocyclopentane-1-carboxylic acid
C6H11NO2
MLLSSTJTARJLHK-UHNVWZDZSA-N
1502035
C1CC(CC1C(=O)O)N

Specifications

(1R, 3S)-3-Aminocyclopentanecarboxylic acid
Authentic
White to Beige
5g
94% min. (HPLC)
1r,3s-3-aminocyclopentanecarboxylic acid, 1r,3s-3-aminocyclopentane-1-carboxylic acid, --1r,3s-3-aminocyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1r,3s, 1r,3s---3-aminocyclopentanecarboxylic acid, cis-3-amino-cyclopentanecarboxylic acid, 1r,3s-3-amino-cyclopentanecarboxylic acid, pubchem18731, 1r,3s---3-amino-cyclopentanecarboxylic acid
C1CC(CC1C(=O)O)N
129.16
129.16
95%,98% ee
97.5% (HPLC) min.
Glass bottle
172.1°C
71830-08-5
C6H11NO2
MLLSSTJTARJLHK-UHNVWZDZSA-N
(1R,3S)-3-aminocyclopentane-1-carboxylic acid
1502035
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (1R,3S)-3-Aminocyclopentanecarboxylic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates