missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-(+)-Camphoric acid, 98+%

Product Code. 11456316
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11456316

Brand: Thermo Scientific Alfa Aesar B23106.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-83-4 | C10H14O4-2 | 198.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-83-4
Molecular Formula C10H14O4-2
Molecular Weight (g/mol) 198.218
MDL Number MFCD00001375
InChI Key LSPHULWDVZXLIL-LDWIPMOCSA-L
Synonym d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
PubChem CID 6918944
IUPAC Name (1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
SMILES CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C

Specifications

Melting Point 183°C to 188°C
Beilstein 2050204
Merck Index 141733
Formula Weight 200.23
Percent Purity ≥98%
Quantity 100 g
Optical Rotation +46° (c=1 in Methanol)
Chemical Name or Material (1R,3S)-(+)-Camphoric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.