missing translation for 'onlineSavingsMsg'
Learn More

2-(1,2-Benzisoxazol-3-yl)acetic acid, 97%, Thermo Scientific™

Catalog Number p-2695953
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
4865-84-3
C9H6NO3
176.15
MFCD02180388
BVSIAYQIMUUCRW-UHFFFAOYSA-M
2-1,2-benzisoxazol-3-yl acetic acid, 1,2-benzisoxazole-3-acetic acid, 1,2-benzisoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl acetic acid, 2-benzo d isoxazol-3-yl acetic acid, 1,2-benzoxazol-3-ylacetic acid, 2-benzo d isoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl ethanoic acid, nvu, zlchem 388
301729
[O-]C(=O)CC1=NOC2=CC=CC=C12
This item is not returnable. View return policy
4865-84-3
C9H6NO3
176.15
MFCD02180388
BVSIAYQIMUUCRW-UHFFFAOYSA-M
2-1,2-benzisoxazol-3-yl acetic acid, 1,2-benzisoxazole-3-acetic acid, 1,2-benzisoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl acetic acid, 2-benzo d isoxazol-3-yl acetic acid, 1,2-benzoxazol-3-ylacetic acid, 2-benzo d isoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl ethanoic acid, nvu, zlchem 388
301729
[O-]C(=O)CC1=NOC2=CC=CC=C12
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 2-(1,2-benzoxazol-3-yl)acetic acid
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 2-(1, 2-Benzisoxazol-3-yl)acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'