Learn More
2,1,3-Benzoxadiazole-5-carboxylic acid, 97%, Maybridge™
Brand: Maybridge CC09701EA
Code : 31
Additional Details : CAS Number : 19155-88-5 Weight : 0.06000kg
Quantity | 10g |
---|
Chemical Identifiers
19155-88-5 | |
164.12 | |
WZUFYJFTOVGJJT-UHFFFAOYSA-N | |
2735454 | |
C1=CC2=NON=C2C=C1C(=O)O |
C7H4N2O3 | |
MFCD00276977 | |
benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid | |
2,1,3-benzoxadiazole-5-carboxylic acid |
Specifications
2, 1, 3-Benzoxadiazole-5-carboxylic acid | |
19155-88-5 | |
100.0 | |
C7H4N2O3 | |
benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid | |
C1=CC2=NON=C2C=C1C(=O)O | |
164.12 | |
164.12 |
10g | |
96.5 | |
97% | |
MFCD00276977 | |
WZUFYJFTOVGJJT-UHFFFAOYSA-N | |
2,1,3-benzoxadiazole-5-carboxylic acid | |
2735454 | |
97% |