Learn More
[2-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Maybridge™
Brand: Maybridge CC34309CB
Code : 31
Additional Details : CAS Number : 61034-86-4 Weight : 0.05025kg
Quantity | 250mg |
---|
Chemical Identifiers
61034-86-4 | |
173.215 | |
PMFMGYSILUCETA-UHFFFAOYSA-N | |
7016493 | |
C1=CC=C(C(=C1)CO)N2C=CC=C2 |
C11H11NO | |
MFCD04115123 | |
2-1h-pyrrol-1-yl phenyl methanol, 2-1h-pyrrol-1-yl benzyl alcohol, 2-pyrrol-1-yl-phenyl-methanol, 2-pyrrol-1-yl phenyl methanol, 2-1h-pyrrol-1-yl benzylalcohol, pubchem11305, 2-pyrrolylphenyl methan-1-ol, 2-pyrrol-1-yl-benzyl alcohol, 1-2-hydroxymethyl phenyl-1h-pyrrole | |
(2-pyrrol-1-ylphenyl)methanol |
Specifications
[2-(1H-Pyrrol-1-yl)phenyl]methanol | |
250mg | |
96.5 | |
97% | |
MFCD04115123 | |
PMFMGYSILUCETA-UHFFFAOYSA-N | |
(2-pyrrol-1-ylphenyl)methanol | |
7016493 | |
97% |
Amber glass bottle | |
61034-86-4 | |
100.0 | |
C11H11NO | |
2-1h-pyrrol-1-yl phenyl methanol, 2-1h-pyrrol-1-yl benzyl alcohol, 2-pyrrol-1-yl-phenyl-methanol, 2-pyrrol-1-yl phenyl methanol, 2-1h-pyrrol-1-yl benzylalcohol, pubchem11305, 2-pyrrolylphenyl methan-1-ol, 2-pyrrol-1-yl-benzyl alcohol, 1-2-hydroxymethyl phenyl-1h-pyrrole | |
C1=CC=C(C(=C1)CO)N2C=CC=C2 | |
173.215 | |
173.21 |