missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl trifluoromethanesulfonate, 97%

Product Code. 15256058 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15256058

Brand: Thermo Scientific Acros 454970010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6226-25-1 | C3H2F6O3S | 232.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6226-25-1
Molecular Formula C3H2F6O3S
Molecular Weight (g/mol) 232.1
InChI Key RTMMSCJWQYWMNK-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyltrifluoromethanesulfonate, 2,2,2-trifluoroethyl triflate, methanesulfonic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluorometanesulfonic acid, 2,2,2-trifluoroethyltrifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluoromethane sulfonate, trifluoroethyltriflate, trifluoroethyl triflate
PubChem CID 80366
IUPAC Name 2,2,2-trifluoroethyl trifluoromethanesulfonate
SMILES C(C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Density 1.6100g/mL
Boiling Point 89.0°C to 91.0°C (740.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula CF3SO3CH2CF3
Quantity 1 g
Specific Gravity 1.61
Formula Weight 232.1
Percent Purity 97%
Chemical Name or Material 2,2,2-Trifluoroethyl trifluoromethanesulfonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,2-Trifluoroethyl trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.