missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2,5-Dimethyl-1,3-thiazol-4-yl)acetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifica
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 171.22 |
Percent Purity | 97% |
Chemical Name or Material | 2-(2, 5-Dimethyl-1, 3-thiazol-4-yl)acetic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'