missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%, ACROS Organics™

Brand:  Acros Organics 168320050

Code : 29

Additional Details : CAS Number : 1118-71-4 Weight : 0.05500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10428920

  • 28.10 € / 5g


EU Stock 2
Add to basket


Description

Description

Acylation reagent
Specifications

Chemical Identifiers

1118-71-4
184.279
dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, pubchem12497
2,2,6,6-tetramethylheptane-3,5-dione
C11H20O2
YRAJNWYBUCUFBD-UHFFFAOYSA-N
70700
CC(C)(C)C(=O)CC(=O)C(C)(C)C

Specifications

2,2,6,6-Tetramethyl-3,5-heptanedione
67°C
Glass bottle
0.88
Colorless to Yellow
1118-71-4
C11H20O2
dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, pubchem12497
YRAJNWYBUCUFBD-UHFFFAOYSA-N
2,2,6,6-tetramethylheptane-3,5-dione
70700
Liquid
0.8800g/mL
Authentic
1.4579 to 1.4599
72.0°C to 73.0°C (6.0mmHg)
5g
98%
(CH3)3CCOCH2COC(CH3)3
Solubility in water: insoluble.
CC(C)(C)C(=O)CC(=O)C(C)(C)C
184.279
184.28
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,2,6,6-Tetramethyl-3,5-heptanedione

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS