Learn More
2-(2-chlorophenoxy)ethanimidamide hydrochloride, 97%, Maybridge™
Brand: Maybridge SPB05621EB
Code : 31
Additional Details : CAS Number : 58403-03-5 Weight : 0.07500kg
Quantity | 25g |
---|
Chemical Identifiers
58403-03-5 | |
221.081 | |
WPWKAEFYVGWTAE-UHFFFAOYSA-N | |
2745914 | |
C1=CC=C(C(=C1)OCC(=N)N)Cl.Cl |
C8H10Cl2N2O | |
MFCD00173805 | |
2-2-chlorophenoxy acetamidine hydrochloride, 2-2-chlorophenoxy ethanimidamide hydrochloride, 2-2-chlorophenoxy acetimidamide hydrochloride, 2-2-chloro-phenoxy-acetamidine hydrochloride, 2-2-chlorophenoxy ethanimidamide monohydrochloride, 2-2-chlorophenoxy acetamide hydrochloride, ethanimidamide, 2-2-chlorophenoxy-, monohydrochloride, 2-2-chlorophenoxy ethanamidine, chloride, 2-2-chlorophenoxy acetamidine hcl, 2-chlorophenoxy ethanimidamide hydrochloride | |
2-(2-chlorophenoxy)ethanimidamide;hydrochloride |
Specifications
2-(2-Chlorophenoxy)ethanimidamide hydrochloride | |
58403-03-5 | |
97% | |
ClC6H4OCH2C(=NH)NH2·HCl | |
2-2-chlorophenoxy acetamidine hydrochloride, 2-2-chlorophenoxy ethanimidamide hydrochloride, 2-2-chlorophenoxy acetimidamide hydrochloride, 2-2-chloro-phenoxy-acetamidine hydrochloride, 2-2-chlorophenoxy ethanimidamide monohydrochloride, 2-2-chlorophenoxy acetamide hydrochloride, ethanimidamide, 2-2-chlorophenoxy-, monohydrochloride, 2-2-chlorophenoxy ethanamidine, chloride, 2-2-chlorophenoxy acetamidine hcl, 2-chlorophenoxy ethanimidamide hydrochloride | |
C1=CC=C(C(=C1)OCC(=N)N)Cl.Cl | |
221.081 | |
221.09 |
25g | |
97.0 | |
C8H10Cl2N2O | |
MFCD00173805 | |
WPWKAEFYVGWTAE-UHFFFAOYSA-N | |
2-(2-chlorophenoxy)ethanimidamide;hydrochloride | |
2745914 | |
97% |