missing translation for 'onlineSavingsMsg'
Learn More

2-(2-chlorophenyl)benzimidazole, 97%, Thermo Scientific™

Catalog Number p-4228532
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3574-96-7
C13H9ClN2
228.68
OILIYWFQRJOPAI-UHFFFAOYSA-N
chlorfenazole, 2-2-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-benzimidazole, chlorfenazole iso, 1h-benzimidazole, 2-2-chlorophenyl, 2-2-chlorophenyl-1h-benzoimidazole, 2-2-chlorophenyl-1h-benzo d imidazole, unii-rt668w2svp, 2-o-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-1,3-benzodiazole
77123
2-(2-chlorophenyl)-1H-benzimidazole
C1=CC=C(C(=C1)C2=NC3=CC=CC=C3N2)Cl
This item is not returnable. View return policy
3574-96-7
C13H9ClN2
228.68
OILIYWFQRJOPAI-UHFFFAOYSA-N
chlorfenazole, 2-2-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-benzimidazole, chlorfenazole iso, 1h-benzimidazole, 2-2-chlorophenyl, 2-2-chlorophenyl-1h-benzoimidazole, 2-2-chlorophenyl-1h-benzo d imidazole, unii-rt668w2svp, 2-o-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-1,3-benzodiazole
77123
2-(2-chlorophenyl)-1H-benzimidazole
C1=CC=C(C(=C1)C2=NC3=CC=CC=C3N2)Cl
CAS Min % 96.0
CAS Max % 100.0
Melting Point 223.2°C to 234.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 228.68
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-chlorophenyl)benzimidazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Chlorophenyl)benzimidazole, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'