missing translation for 'onlineSavingsMsg'
Learn More

2(2-Ethoxyethoxy)ethanol, 98+%, Thermo Scientific Chemicals

Catalog Number p-3778525
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
This item is not returnable. View return policy
111-90-0
MFCD00002872
XXJWXESWEXIICW-UHFFFAOYSA-N
diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
8146
CHEBI:40572
2-(2-ethoxyethoxy)ethanol
CCOCCOCCO
This item is not returnable. View return policy
111-90-0
MFCD00002872
XXJWXESWEXIICW-UHFFFAOYSA-N
diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
8146
CHEBI:40572
2-(2-ethoxyethoxy)ethanol
CCOCCOCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -80.0°C
Color Colorless
Density 0.9900g/mL
Boiling Point 197.0°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Molecular Formula C6H14O3
Linear Formula C2H5OCH2CH2OCH2CH2OH
Refractive Index 1.4260 to 1.4280
Quantity 1 L
Beilstein 01, II, 520
Merck Index 15, 3134
Specific Gravity 0.99
Molecular Weight (g/mol) 134.175
Formula Weight 134.17
Percent Purity ≥98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2(2-Ethoxyethoxy)ethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'