missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanediol, 98%, mixture of racemic and meso forms, techn., Thermo Scientific Chemicals

Catalog Number p-4509393
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
513-85-9
C4H10O2
90.12
OWBTYPJTUOEWEK-UHFFFAOYSA-N
2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-.
262
CHEBI:62064
butane-2,3-diol
CC(C(C)O)O
513-85-9
C4H10O2
90.12
OWBTYPJTUOEWEK-UHFFFAOYSA-N
2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-.
262
CHEBI:62064
butane-2,3-diol
CC(C(C)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 19.0°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 180.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH(OH)CH3
Refractive Index 1.4330 to 1.4370
Quantity 250 mL
Beilstein 01,479
Merck Index 15,1571
Specific Gravity 1.01
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol, ether and diisopropyl ether
Formula Weight 90.12
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 2, 3-Butanediol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'