missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione, 99%, Thermo Scientific Chemicals

Catalog Number p-78180
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
431-03-8
C4H6O2
86.09
MFCD00008756
QSJXEFYPDANLFS-UHFFFAOYSA-N
2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
650
CHEBI:16583
butane-2,3-dione
CC(=O)C(=O)C
This item is not returnable. View return policy
431-03-8
C4H6O2
86.09
MFCD00008756
QSJXEFYPDANLFS-UHFFFAOYSA-N
2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
650
CHEBI:16583
butane-2,3-dione
CC(=O)C(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -4°C to 2°C
Color Yellow
Density 0.9850g/mL
Boiling Point 88.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCOCH3
Refractive Index 1.3910 to 1.3990
Quantity 5 mL
Beilstein 01, 769
Fieser 07,21
Merck Index 15, 2969
Specific Gravity 0.985
Solubility Information Solubility in water: 200g/L (20°C). Other solubilities: miscible with alcohol, ether, very soluble in acetone, soluble in oils, fats, propylene glycol, glycerol, soluble in carbitols, all important organic solven, ts
Formula Weight 86.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2,3-Butanedione, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'