missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloroanisole, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
Description
Synthesis Material Intermediates. The product has been used to develop an analytical method for the determination of biogenic halogenated anisoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Synthesis Material Intermediates. The product has been used to develop an analytical method for the determination of biogenic halogenated anisoles.
Solubility
Soluble in water (86.9 mg/L).
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, strong acids. Keep container tightly closed.
Synthesis Material Intermediates. The product has been used to develop an analytical method for the determination of biogenic halogenated anisoles.
Solubility
Soluble in water (86.9 mg/L).
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, strong acids. Keep container tightly closed.
Specifications
Melting Point | 31°C to 33°C |
Boiling Point | 115°C to 116°C (12 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 50 g |
Beilstein | 1909166 |
Solubility Information | Soluble in water (86.9mg/L). |
Formula Weight | 177.03 |
Percent Purity | 97% |
Chemical Name or Material | 2,3-Dichloroanisole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'