missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number 10026233
95.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1521-38-6
C9H10O4
182.18
MFCD00002432
FODBVCSYJKNBLO-UHFFFAOYSA-N
o-veratric acid, benzoic acid, 2,3-dimethoxy, benzoic acid, dimethoxy, 2,3-dimethoxy benzoic acid, dimethoxybenzoic acid,2,3, o-veratric acid 6ci,7ci,8ci, 2,3-dimethoxybenzoicacid, dimethoxybenzoic acid, pubchem4948, 2,3-dimethoxy-benzoicaci
15204
2,3-dimethoxybenzoic acid
COC1=CC=CC(C(O)=O)=C1OC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1521-38-6
C9H10O4
182.18
MFCD00002432
FODBVCSYJKNBLO-UHFFFAOYSA-N
o-veratric acid, benzoic acid, 2,3-dimethoxy, benzoic acid, dimethoxy, 2,3-dimethoxy benzoic acid, dimethoxybenzoic acid,2,3, o-veratric acid 6ci,7ci,8ci, 2,3-dimethoxybenzoicacid, dimethoxybenzoic acid, pubchem4948, 2,3-dimethoxy-benzoicaci
15204
2,3-dimethoxybenzoic acid
COC1=CC=CC(C(O)=O)=C1OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 121°C to 124°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula (CH3O)2C6H3CO2H
Quantity 100 g
Beilstein 10, I, 174
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 182.18
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2, 3-Dimethoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dimethoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'