Learn More
2-(3-Pyridyl)ethan-1-ol, Maybridge™
Brand: Maybridge KM09875DA
Code : 31
Additional Details : CAS Number : 6293-56-7 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
6293-56-7 | |
123.155 | |
YPWSASPSYAWQRK-UHFFFAOYSA-N | |
80515 | |
C1=CC(=CN=C1)CCO |
C7H9NO | |
MFCD00831047 | |
3-pyridineethanol, 3-2-hydroxyethyl pyridine, 2-pyridin-3-yl ethanol, 2-3-pyridyl ethan-1-ol, 2-3-pyridyl-1-ethanol, 2-pyridin-3-yl ethan-1-ol, 3-pyridylethanol, 2-3-pyridyl ethanol, pyridyl-3-methylcarbinol, 3-hydroxyethyl pyridine | |
2-pyridin-3-ylethanol |
Specifications
2-(3-Pyridyl)ethan-1-ol | |
1g | |
97.0 | |
C7H9NO | |
3-pyridineethanol, 3-2-hydroxyethyl pyridine, 2-pyridin-3-yl ethanol, 2-3-pyridyl ethan-1-ol, 2-3-pyridyl-1-ethanol, 2-pyridin-3-yl ethan-1-ol, 3-pyridylethanol, 2-3-pyridyl ethanol, pyridyl-3-methylcarbinol, 3-hydroxyethyl pyridine | |
C1=CC(=CN=C1)CCO | |
123.155 | |
123.15 |
Amber glass bottle | |
6293-56-7 | |
97% | |
MFCD00831047 | |
YPWSASPSYAWQRK-UHFFFAOYSA-N | |
2-pyridin-3-ylethanol | |
80515 | |
97% |