missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzoyl chloride, 99%, ACROS Organics™

Brand:  Acros Organics 309330050

Code : 29

Additional Details : CAS Number : 88419-56-1 Weight : 0.05500kg Transport : UN number : 3265 Chem class : 8 Pack group : III

Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10006013

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.
View alternative products »


Specifications

Chemical Identifiers

2, 4, 5-Trifluorobenzoyl chloride
98.5
≥98.5%
C7H2ClF3O
MFCD00061204
88419-56-1
100.0
99%
F3C6H2COCl

Specifications

2, 4, 5-Trifluorobenzoyl chloride
194.54
5g
98.5
Authentic
1.4970 to 1.4990 (20°C, 589nm)
≥98.5%
C7H2ClF3O
MFCD00061204
C1=C(C(=CC(=C1F)F)F)C(=O)Cl
145164
1.5200g/mL
Clear colorless
88419-56-1
100.0
Glass bottle
Liquid
99%
F3C6H2COCl
STBGCAUUOPNJBH-UHFFFAOYSA-N
194.537
Videos
SDS