missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorophenylboronic acid, 97%, ACROS Organics™

Brand:  Acros Organics 393800250

Code : 29

Additional Details : CAS Number : 247564-72-3 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 11935791

  • 371.00 € / 25g

Estimated Shipment: 13-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

247564-72-3
175.9
2,4,5-trifluorophenyl boronic acid, 2,4,5-trifluorobenzeneboronic acid, boronic acid, b-2,4,5-trifluorophenyl, boronic acid, 2,4,5-trifluorophenyl, 2,4,5-trifluorophenylboronicacid, pubchem1787, acmc-1cd74
(2,4,5-trifluorophenyl)boronic acid
C6H4BF3O2
KCHHKNCSISEAAE-UHFFFAOYSA-N
2783133
B(C1=CC(=C(C=C1F)F)F)(O)O

Specifications

2, 4, 5-Trifluorophenylboronic acid
118.0°C to 119.0°C
247564-72-3
C6H4BF3O2
2,4,5-trifluorophenyl boronic acid, 2,4,5-trifluorobenzeneboronic acid, boronic acid, b-2,4,5-trifluorophenyl, boronic acid, 2,4,5-trifluorophenyl, 2,4,5-trifluorophenylboronicacid, pubchem1787, acmc-1cd74
KCHHKNCSISEAAE-UHFFFAOYSA-N
(2,4,5-trifluorophenyl)boronic acid
2783133
97%
Glass bottle
25g
96% min. (HPLC)
F3C6H2B(OH)2
Solubility in water: .. Other solubilities: soluble in methanol
B(C1=CC(=C(C=C1F)F)F)(O)O
175.9
175.9
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,4,5-Trifluorophenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates