missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-hydroxy-5-nitrosopyrimidine, 97%, Thermo Scientific™

Catalog Number 11489412
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
2387-48-6
C4H5N5O2
155.117
MFCD00023263
HVMRLFSFHWCUCG-UHFFFAOYSA-N
2,4-diamino-6-hydroxy-5-nitrosopyrimidine, 2,6-diamino-5-nitrosopyrimidin-4-ol, 4-pyrimidinol, 2,6-diamino-5-nitroso, 4 1h-pyrimidinone, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitroso-4-pyrimidinol, 4-pyrimidol, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitrosopyrimidin-4 3h-one, d04tlb, acmc-1cg71, 4-pyrimidol,6-diamino-5-nitroso
75436
2,6-diamino-5-nitroso-1H-pyrimidin-4-one
C1(=C(NC(=NC1=O)N)N)N=O
This item is not returnable. View return policy
2387-48-6
C4H5N5O2
155.117
MFCD00023263
HVMRLFSFHWCUCG-UHFFFAOYSA-N
2,4-diamino-6-hydroxy-5-nitrosopyrimidine, 2,6-diamino-5-nitrosopyrimidin-4-ol, 4-pyrimidinol, 2,6-diamino-5-nitroso, 4 1h-pyrimidinone, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitroso-4-pyrimidinol, 4-pyrimidol, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitrosopyrimidin-4 3h-one, d04tlb, acmc-1cg71, 4-pyrimidol,6-diamino-5-nitroso
75436
2,6-diamino-5-nitroso-1H-pyrimidin-4-one
C1(=C(NC(=NC1=O)N)N)N=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >360°C
Quantity 25 g
Formula Weight 155.12
Percent Purity 97%
Chemical Name or Material 2,4-Diamino-6-hydroxy-5-nitrosopyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'