missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-fluoropyrimidine, 97%, ACROS Organics™

Brand:  Acros Organics 443220050

Code : 29

Additional Details : CAS Number : 2927-71-1 Weight : 0.05500kg Transport : UN number : 3261 Chem class : 8 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 11946561

  • 440.00 € / 5g

Estimated Shipment: 07-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2927-71-1
166.96
WHPFEQUEHBULBW-UHFFFAOYSA-N
250705
FC1=CN=C(Cl)N=C1Cl
C4HCl2FN2
MFCD00233551
2,4-dichloro-5-fluoro-pyrimidine, 5-fluoro-2,4-dichloropyrimidine, 2,6-dichloro-5-fluororacil, 2,4-dichloro-5-fluoro pyrimidine, pyrimidine, 2,4-dichloro-5-fluoro, 2,6-dichloro-5-fluoropyrimidine, 2,4-dichlor-5-fluorpyrimidin, pubchem5290, ksc204s2l, 2,4-dichloro-5fluoropyrimidine
2,4-dichloro-5-fluoropyrimidine

Specifications

2, 4-Dichloro-5-fluoropyrimidine
Glass Bottle
Brown to White
5g
96.0
96% min. (GC)
MFCD00233551
2,4-dichloro-5-fluoro-pyrimidine, 5-fluoro-2,4-dichloropyrimidine, 2,6-dichloro-5-fluororacil, 2,4-dichloro-5-fluoro pyrimidine, pyrimidine, 2,4-dichloro-5-fluoro, 2,6-dichloro-5-fluoropyrimidine, 2,4-dichlor-5-fluorpyrimidin, pubchem5290, ksc204s2l, 2,4-dichloro-5fluoropyrimidine
FC1=CN=C(Cl)N=C1Cl
166.96
166.97
97%
Authentic
67°C to 68°C (6.0 Torr)
38°C to 40°C
2927-71-1
100.0
C4HCl2FN2
3261
WHPFEQUEHBULBW-UHFFFAOYSA-N
2,4-dichloro-5-fluoropyrimidine
250705
Powder or Crystalline Powder or Low Melting Solid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,4-Dichloro-5-fluoropyrimidine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates