missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenyl isothiocyanate, 95%, Thermo Scientific™

Catalog Number p-4258971
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
33904-03-9
C9H9NO2S
195.24
CNXSCEPTSXPBTP-UHFFFAOYSA-N
2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
2736202
1-isothiocyanato-2,4-dimethoxybenzene
COC1=CC(=C(C=C1)N=C=S)OC
This item is not returnable. View return policy
33904-03-9
C9H9NO2S
195.24
CNXSCEPTSXPBTP-UHFFFAOYSA-N
2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
2736202
1-isothiocyanato-2,4-dimethoxybenzene
COC1=CC(=C(C=C1)N=C=S)OC
CAS Min % 94.0
CAS Max % 100.0
Melting Point 54.0°C to 55.0°C
Color Yellow
Boiling Point 166.0°C to 167.0°C (6.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3O)2C6H3NCS
Quantity 1 g
Formula Weight 195.24
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2, 4-Dimethoxyphenyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethoxyphenyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'