missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxypyrimidine-5-boronic acid, 95%, Thermo Scientific Chemicals

Catalog Number 11936141
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
89641-18-9
C6H9BN2O4
183.96
LKGKUACPLXCVOF-UHFFFAOYSA-N
2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-yl boronic acid, 2,4-dimethoxyprimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-ylboronic acid, 2,4-dimethoxy-5-pyrimidineboronic acid, 2,4-dimethoxypryimidine-5-boronic acid, 2,4-dimethoxy-5-pyrimidinylboronic acid, 5-borono-2,3-dimethoxypyrimidine, 2,4-dimethoxypyrimidine-5-boronicacid
2736209
This item is not returnable. View return policy
89641-18-9
C6H9BN2O4
183.96
LKGKUACPLXCVOF-UHFFFAOYSA-N
2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-yl boronic acid, 2,4-dimethoxyprimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-ylboronic acid, 2,4-dimethoxy-5-pyrimidineboronic acid, 2,4-dimethoxypryimidine-5-boronic acid, 2,4-dimethoxy-5-pyrimidinylboronic acid, 5-borono-2,3-dimethoxypyrimidine, 2,4-dimethoxypyrimidine-5-boronicacid
2736209

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 107°C to 109°C
Color White to Yellow
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 183.96
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 4-Dimethoxypyrimidine-5-boronic acid
compliance-icons
Product Identifier
  • 2,4-Dimethoxypyrimidine-5-boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'