missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,5-Dibromotoluene, 98%

27.50 € - 106.00 €

Chemical Identifiers

CAS 615-59-8
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000090
InChI Key QKEZTJYRBHOKHH-UHFFFAOYSA-N
Synonym 2,5-dibromotoluene, benzene, 1,4-dibromo-2-methyl, toluene, 2,5-dibromo, 2,5-dibromo toluene, 3,6-dibromotoluene, 1,4-dibromo-2-methyl-benzene, pubchem3922, acmc-209mu2, chemwish ic06904
PubChem CID 12006
IUPAC Name 1,4-dibromo-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11444017 SDS View Documents Alfa Aesar™
A19418.06
5g 27.50 €
5g View alternative products
Save Up to 
»
EU Stock 9
Please to purchase this item. Need a web account? Register with us today!
11454017 SDS View Documents Alfa Aesar™
A19418.14
25g 106.00 €
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-09-2019
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

615-59-8
249.933
QKEZTJYRBHOKHH-UHFFFAOYSA-N
12006
CC1=C(C=CC(=C1)Br)Br
C7H6Br2
MFCD00000090
2,5-dibromotoluene, benzene, 1,4-dibromo-2-methyl, toluene, 2,5-dibromo, 2,5-dibromo toluene, 3,6-dibromotoluene, 1,4-dibromo-2-methyl-benzene, pubchem3922, acmc-209mu2, chemwish ic06904
1,4-dibromo-2-methylbenzene

Specifications

2,5-Dibromotoluene
Aromatic
5°C to 6°C
615-59-8
MFCD00000090
2,5-dibromotoluene, benzene, 1,4-dibromo-2-methyl, toluene, 2,5-dibromo, 2,5-dibromo toluene, 3,6-dibromotoluene, 1,4-dibromo-2-methyl-benzene, pubchem3922, acmc-209mu2, chemwish ic06904
CC1=C(C=CC(=C1)Br)Br
249.933
249.94
>110°C (230°F)
106°C to 108°C (10mmHg)
5g
C7H6Br2
1859123
QKEZTJYRBHOKHH-UHFFFAOYSA-N
1,4-dibromo-2-methylbenzene
12006
98%