missing translation for 'onlineSavingsMsg'
Learn More

2(5H)-Furanone, 95%, Thermo Scientific Chemicals

Catalog Number p-4258871
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
497-23-4
C4H4O2
84.07
MFCD00005376
VIHAEDVKXSOUAT-UHFFFAOYSA-N
2 5h-furanone, furan-2 5h-one, butenolide, 2-butenolide, gamma-crotonolactone, 2-buten-4-olide, isocrotonolactone, 2-oxo-2,5-dihydrofuran, crotonolactone, 2-5h-furanone
10341
CHEBI:38118
2H-furan-5-one
O=C1OCC=C1
This item is not returnable. View return policy
497-23-4
C4H4O2
84.07
MFCD00005376
VIHAEDVKXSOUAT-UHFFFAOYSA-N
2 5h-furanone, furan-2 5h-one, butenolide, 2-butenolide, gamma-crotonolactone, 2-buten-4-olide, isocrotonolactone, 2-oxo-2,5-dihydrofuran, crotonolactone, 2-5h-furanone
10341
CHEBI:38118
2H-furan-5-one
O=C1OCC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 3.0°C to 4.0°C
Color Amber or Colorless to Yellow
Density 1.1900g/mL
Boiling Point 212.0°C to 214.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.19
Solubility Information Solubility in water: insoluble
Formula Weight 84.07
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2(5H)-Furanone
Show More Show Less
compliance-icons
Product Identifier
  • 2-(5H)-Furanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'