missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol sodium salt hydrate, Honeywell Fluka™

Product Code. p-7072190 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15635120

Brand: Honeywell-Fluka 361801G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥97.0% (calc. based on dry substance, AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
PubChem CID 23697355
ChEBI CHEBI:948
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Quantity 1 g
UN Number NONH for all modes of transport
Beilstein 3641229
Formula Weight 290.08g/mol (anhydrous basis)
Percent Purity ≥97% (calculated based on dry substance, AT)
Chemical Name or Material 2,6-Dichloroindophenol sodium salt hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.