missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Difluoro-N-(5-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59377.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15414996

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2,6-Difluoro-N-(5-fluoro-2-methylphenyl)benzamide
265.23
MFCD04804765

Specifications

2,6-Difluoro-N-(5-fluoro-2-methylphenyl)benzamide
Solid
265.23
1g
97%
MFCD04804765
Videos
SDS