Learn More
2,6-Dimethylbenzonitrile, 99%, Acros Organics™
Brand: Acros Organics 355590250
Code : NEW
Additional Details : CAS Number : 6575-13-9 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
6575-13-9 | |
131.18 | |
benzonitrile, 2,6-dimethyl, unii-1zh37s5b0z, 2,6-dimethylbenzenecarbonitrile, 2,6-dimethyl-benzonitrile, 2-cyano-m-xylene, ksc494c5t, 2,6-dimethylbenz0nitrile, 3-cyano-2,4-dimethylphenyl | |
2,6-dimethylbenzonitrile |
C9H9N | |
QSACPWSIIRFHHR-UHFFFAOYSA-N | |
81057 | |
CC1=C(C(=CC=C1)C)C#N |
Specifications
2,6-Dimethylbenzonitrile | |
88.0°C to 92.0°C | |
6575-13-9 | |
C9H9N | |
QSACPWSIIRFHHR-UHFFFAOYSA-N | |
2,6-dimethylbenzonitrile | |
81057 | |
99% |
Authentic | |
25g | |
98.5% min. (GC) | |
benzonitrile, 2,6-dimethyl, unii-1zh37s5b0z, 2,6-dimethylbenzenecarbonitrile, 2,6-dimethyl-benzonitrile, 2-cyano-m-xylene, ksc494c5t, 2,6-dimethylbenz0nitrile, 3-cyano-2,4-dimethylphenyl | |
CC1=C(C(=CC=C1)C)C#N | |
131.18 | |
131.18 |
Safety and Handling
- 2,6-Dimethylbenzonitrile
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.