Learn More
Julabo™ 2-Adapters for Presto™ Temperature Control System
Brand:  Julabo™ 8890057
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15234263
  • 304.00 € / Pack of 2
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Specifications
Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System
2-Adapters
M24 x 1.5 Female to NPT 1 in. Female