Learn More
Julabo™ 2-Adapters for Presto™ Temperature Control System
Brand: Julabo™ 8890058
Code : NEW
Additional Details : Weight : 0.50000kg
Specifications
Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System | |
2-Adapters | |
M24 x 1.5 Female to NPT 1/4 in. Male |