Learn More
Julabo™ 2-Adapters for Recirculating Cooler
Brand: Julabo™ 8890047
Code : A0
Additional Details : Weight : 0.50000kg
Specifications
Suitable for FL1203, FL1703, FL2503, FL4003 Recirculating Cooler | |
2-Adapters | |
G 3/4 in. Female to NPT 1/2 in. Male |