Learn More
Julabo™ 2-Adapters for Recirculating Cooler
Brand:  Julabo™ 8890040
Code : A0
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11914708
  • 133.00 € / Pack of 2
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Specifications
Suitable for FL1203, FL1703, FL2503, FL4003 Recirculating Cooler
2-Adapters
G 3/4 in. Female to M16 x 1 Male