Learn More
Julabo™ 2-Adapters for Recirculating Cooler
Brand:  Julabo™ 8890051
Code : A0
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11924718
  • 205.00 € / Pack of 2
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
Specifications
Suitable for FL11006, FL20006, FL2506, FL4006, FL7006 Recirculating Cooler
2-Adapters
G 1 1/4 in. Female to NPT 1 in. Male