Learn More
Julabo™ 2-Adapters for Recirculating Cooler
Brand: Julabo™ 8890045
Code : A0
Additional Details : Weight : 0.50000kg
Specifications
Suitable for FL11006, FL20006, FL2506, FL4006, FL7006 Recirculating Cooler | |
2-Adapters | |
G 1 1/4 in. Female to 3/4 in. Barbed Fitting |