missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-benzyloxypyridine, 98.5%, ACROS Organics™

Brand:  Acros Organics 162980250

Code : 29

Additional Details : CAS Number : 24016-03-3 Weight : 0.07500kg

Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10428890

  • 105.30 € / 25g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

24016-03-3
200.241
NMCBWICNRJLKKM-UHFFFAOYSA-N
C12H12N2O
MFCD00006316
90334

Specifications

2-Amino-3-benzyloxypyridine
24016-03-3
100.0
Glass bottle
98.5%
C12H12N2O
MFCD00006316
NMCBWICNRJLKKM-UHFFFAOYSA-N
3-phenylmethoxypyridin-2-amine
90334
Crystalline Powder
98.5%
98.0
Authentic
≥98.0%
94°C to 98°C
Brown to Green
25g
C1=CC=C(C=C1)COC2=C(N=CC=C2)N
200.241
200.24
Videos
SDS