missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog Number 10001730
31.46 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16867-03-1
C5H6N2O
110.12
MFCD00006317
BMTSZVZQNMNPCT-UHFFFAOYSA-N
2-amino-3-hydroxypyridine, 2-amino-3-pyridinol, 3-pyridinol, 2-amino, 2-amino-3-pyridol, 3-hydroxy-2-pyridinamine, 2-amino-pyridin-3-ol, 2-amino-3-hydroxy pyridine, unii-p8740pew4b, 3-hydroxy-2-aminopyridine, pubchem2574
28114
2-aminopyridin-3-ol
C1=CC(=C(N=C1)N)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16867-03-1
C5H6N2O
110.12
MFCD00006317
BMTSZVZQNMNPCT-UHFFFAOYSA-N
2-amino-3-hydroxypyridine, 2-amino-3-pyridinol, 3-pyridinol, 2-amino, 2-amino-3-pyridol, 3-hydroxy-2-pyridinamine, 2-amino-pyridin-3-ol, 2-amino-3-hydroxy pyridine, unii-p8740pew4b, 3-hydroxy-2-aminopyridine, pubchem2574
28114
2-aminopyridin-3-ol
C1=CC(=C(N=C1)N)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 170°C to 176°C
Color Beige-Gray to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HO(C5H3N)NH2
Quantity 25 g
Beilstein 22, II, 408
Formula Weight 110.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-hydroxypyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-hydroxypyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'