missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-iodopyridine, 97%, Thermo Scientific Chemicals

Catalog Number 11926411
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
104830-06-0
C5H5IN2
220.01
UUDNBWSHTUFGDQ-UHFFFAOYSA-N
2-amino-3-iodopyridine, 3-iodopyrid-2-ylamine, 3-iodo-pyridin-2-ylamine, 3-iodo-2-pyridylamine, 2-pyridinamine, 3-iodo, iodo-aminopyridine, pubchem1265, 2-amino-3-iodpyridine, acmc-20a0df
2763151
3-iodopyridin-2-amine
C1=CC(=C(N=C1)N)I
This item is not returnable. View return policy
104830-06-0
C5H5IN2
220.01
UUDNBWSHTUFGDQ-UHFFFAOYSA-N
2-amino-3-iodopyridine, 3-iodopyrid-2-ylamine, 3-iodo-pyridin-2-ylamine, 3-iodo-2-pyridylamine, 2-pyridinamine, 3-iodo, iodo-aminopyridine, pubchem1265, 2-amino-3-iodpyridine, acmc-20a0df
2763151
3-iodopyridin-2-amine
C1=CC(=C(N=C1)N)I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 89.6°C to 90.6°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 220.01
Percent Purity 97%
Physical Form Crystalline Powder, Powder or Needles
Chemical Name or Material 2-Amino-3-iodopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-iodopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'