missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-pyridyl)thiazole, 97%, Thermo Scientific Chemicals

Catalog Number 15474798
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
30235-26-8
C8H7N3S
177.225
MFCD00460417
BLKHMTAXNXLDJP-UHFFFAOYSA-N
4-pyridin-2-yl thiazol-2-amine, 4-pyridin-2-yl-thiazol-2-ylamine, 4-pyridin-2-yl-1,3-thiazol-2-amine, 2-amino-4-2-pyridyl thiazole, 2-thiazolamine, 4-2-pyridinyl, 4-2-pyridinyl-1,3-thiazol-2-amine, 4-2-pyridinyl-2-thiazolamine, 4-2-pyridinyl thiazol-2-amine, 4-2-pyridyl-1,3-thiazole-2-ylamine, 4-pyridin-2-yl-thiazole-2-ylamine
1092459
4-pyridin-2-yl-1,3-thiazol-2-amine
C1=CC=NC(=C1)C2=CSC(=N2)N
This item is not returnable. View return policy
30235-26-8
C8H7N3S
177.225
MFCD00460417
BLKHMTAXNXLDJP-UHFFFAOYSA-N
4-pyridin-2-yl thiazol-2-amine, 4-pyridin-2-yl-thiazol-2-ylamine, 4-pyridin-2-yl-1,3-thiazol-2-amine, 2-amino-4-2-pyridyl thiazole, 2-thiazolamine, 4-2-pyridinyl, 4-2-pyridinyl-1,3-thiazol-2-amine, 4-2-pyridinyl-2-thiazolamine, 4-2-pyridinyl thiazol-2-amine, 4-2-pyridyl-1,3-thiazole-2-ylamine, 4-pyridin-2-yl-thiazole-2-ylamine
1092459
4-pyridin-2-yl-1,3-thiazol-2-amine
C1=CC=NC(=C1)C2=CSC(=N2)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Quantity 10 g
Formula Weight 177.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-4-(2-pyridyl)thiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'