Learn More
2-Amino-5-chlorobenzoxazole, 97%, ACROS Organics™
Brand: Acros Organics 103380250
Code : 29
Additional Details : CAS Number : 61-80-3 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 25g |
Chemical Identifiers
61-80-3 | |
168.58 | |
YGCODSQDUUUKIV-UHFFFAOYSA-N | |
6103 | |
5-chloro-1,3-benzoxazol-2-amine |
C7H5ClN2O | |
MFCD00005770 | |
zoxazolamine, 2-amino-5-chlorobenzoxazole, 5-chlorobenzo d oxazol-2-amine, flexin, 2-benzoxazolamine, 5-chloro, contrazole, deflexol, flexilon, zoxamin, zoxine | |
CHEBI:35053 | |
C1=CC2=C(C=C1Cl)N=C(O2)N |
Specifications
2-Amino-5-chlorobenzoxazole | |
Glass bottle | |
180.0°C to 185.0°C | |
97% | |
96.0 | |
97% | |
MFCD00005770 | |
zoxazolamine, 2-amino-5-chlorobenzoxazole, 5-chlorobenzo d oxazol-2-amine, flexin, 2-benzoxazolamine, 5-chloro, contrazole, deflexol, flexilon, zoxamin, zoxine | |
C1=CC2=C(C=C1Cl)N=C(O2)N | |
168.58 | |
CHEBI:35053 | |
Powder |
Authentic | |
Brown to Brown-Yellow | |
25g | |
61-80-3 | |
100.0 | |
C7H5ClN2O | |
15, 10399 | |
YGCODSQDUUUKIV-UHFFFAOYSA-N | |
5-chloro-1,3-benzoxazol-2-amine | |
6103 | |
168.58 | |
97% |
Safety and Handling
- 2-Amino-5-chlorobenzoxazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
Hazard Statement
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.