missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylpyridine, 95%, Thermo Scientific Chemicals

Catalog Number p-2697946
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
3430-17-9
C6H6BrN
172.03
MFCD00239380
PZSISEFPCYMBDL-UHFFFAOYSA-N
2-bromo-3-picoline, 2-bromo-3-methylpyridin, 2-bromo-3-methyl pyridine, 2-bromo-3-methyl-pyridine, 3-methyl-2-bromopyridine, pyridine, 2-bromo-3-methyl, 2-brom-3-methylpyridin, pubchem9208, 2-bromo-3-mehtylpyridine
220832
2-bromo-3-methylpyridine
CC1=CC=CN=C1Br
This item is not returnable. View return policy
3430-17-9
C6H6BrN
172.03
MFCD00239380
PZSISEFPCYMBDL-UHFFFAOYSA-N
2-bromo-3-picoline, 2-bromo-3-methylpyridin, 2-bromo-3-methyl pyridine, 2-bromo-3-methyl-pyridine, 3-methyl-2-bromopyridine, pyridine, 2-bromo-3-methyl, 2-brom-3-methylpyridin, pubchem9208, 2-bromo-3-mehtylpyridine
220832
2-bromo-3-methylpyridine
CC1=CC=CN=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.5440g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5650 to 1.57
Quantity 5 mL
Specific Gravity 1.544
Formula Weight 172.02
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-methylpyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-3-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'