missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7055910
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
476620-54-9
C7H3BrF2O
221.001
MFCD16659662
ZHSZEWFWVDLMDL-UHFFFAOYSA-N
benzaldehyde, 2-bromo-4,5-difluoro, acmc-209ka1, 2-bromo-4,5-difluoro-benzaldehyde
21867425
2-bromo-4,5-difluorobenzaldehyde
C1=C(C(=CC(=C1F)F)Br)C=O
This item is not returnable. View return policy
476620-54-9
C7H3BrF2O
221.001
MFCD16659662
ZHSZEWFWVDLMDL-UHFFFAOYSA-N
benzaldehyde, 2-bromo-4,5-difluoro, acmc-209ka1, 2-bromo-4,5-difluoro-benzaldehyde
21867425
2-bromo-4,5-difluorobenzaldehyde
C1=C(C(=CC(=C1F)F)Br)C=O

It is an important intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Beilstein 20365202
Solubility Information Slightly soluble in water.
Formula Weight 221
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Bromo-4,5-difluorobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'